Learn More
RAB31 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15485-1-AP-20UL
This item is not returnable.
View return policy
Description
Small GTP-binding proteins of the RAB family, such as RAB31, play essential roles in vesicle and granule targeting.[supplied by OMIM, Jul 2009].Specifications
| RAB31 | |
| Polyclonal | |
| Unconjugated | |
| RAB31 | |
| Rab22B, RAB31, Ras related protein Rab 22B, Ras related protein Rab 31 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11031 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13636 | |
| RAB31 | |
| RAB31 Fusion Protein Ag7833 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |