missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB30 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16328-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
RAB30 (Ras-related protein Rab-30) encodes for small GTPases Rab which are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. Required for maintaining the structural integrity of the Golgi apparatus, possibly by mediating interactions with cytoplasmic scaffolding proteins.Specifications
RAB30 | |
Polyclonal | |
Unconjugated | |
RAB30 | |
RAB30, Ras related protein Rab 30 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27314, 308821 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15771 | |
Rab30 | |
RAB30 Fusion Protein Ag9444 | |
20 μL | |
Primary | |
Rat, Human | |
Antibody | |
IgG |