Learn More
RAB27A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17817-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified.Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.Specifications
| RAB27A | |
| Polyclonal | |
| Unconjugated | |
| RAB27A | |
| GS2, GTP binding protein Ram, HsT18676, Rab 27, RAB27, RAB27A, RAM, Ras related protein Rab 27A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5873 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51159 | |
| RAB27A | |
| RAB27A Fusion Protein Ag12136 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |