missing translation for 'onlineSavingsMsg'
Learn More

RAB1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17824-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887814

  • 3468.15 NOK / 150µL
Estimated Shipment: 13-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB1B regulates vesicular transport between the endoplasmic reticulum and successive Golgi compartments. Plays a role in the initial events of the autophagic vacuole development which take place at specialized regions of the endoplasmic reticulum.
Specifications

Specifications

RAB1B
Polyclonal
Unconjugated
RAB1B
RAB1B, Ras related protein Rab 1B
Rabbit
Antigen Affinity Chromatography
RUO
76308, 81876
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D1G1, Q9H0U4
Rab1b
RAB1B Fusion Protein Ag12197
150 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers