missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11671-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.Specifications
RAB1A | |
Polyclonal | |
Unconjugated | |
RAB1A | |
RAB1, RAB1A, Ras related protein Rab 1A, YPT1, YPT1 related protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19324, 5861, 81754 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62820, P62821, Q6NYB7 | |
RAB1A | |
RAB1A Fusion Protein Ag2264 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |