missing translation for 'onlineSavingsMsg'
Learn More

RAB11A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20229-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16899744

  • 1611.84 NOK / 20µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to RAB11A, and has no cross reaction to RAB11B.

Rab11A is a member of the YPT-1 subfamily of Ras-related GTP-binding proteins. Rab11A mRNA has been detected in a variety of tissues including brain, testis, spleen, heart, and gastrointestinal mucosa. In the gastric fundus the Rab11A protein is expressed in parietal cell tubulovesicles membranes. Expression of the Rab11A protein appears to occur in discrete epithelial cell populations where it localizes to apical vesicular populations. Recent studies have shown Rab11A is essential for resecretion of alpha-synuclein out of neuronal cells via endosome recycling and is required for exocytosis of discoidal/fusiform vesicles of bladder umbrella cells. Rab11A is also implicated in biogenesis of Birbeck granules and endosomal activation of the PI3K/AKT signaling pathway via G beta gamma trafficking.
Specifications

Specifications

RAB11A
Polyclonal
Unconjugated
RAB11A
Rab 11, RAB11, RAB11A, Ras related protein Rab 11A, YL8
Rabbit
Antigen Affinity Chromatography
RUO
53869, 8766
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62491, P62492
Rab11a
Peptide
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers