Learn More
Epredia™ RA Lamb BlockStore Storage Options for the LabStack Storage System
Product Code. 12181830 Shop All Epredia Products
Change view
Click to view available options
Product Type:
8-Drawer Cabinet
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Color Unit Size
12181830 8-Drawer Cabinet Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12181830 Supplier Epredia™ Supplier No. E43/22GREY
  • ONLINE EXCLUSIVE PRICE
    4978.61 NOK
    4501.49 NOK / Pack of 1
    Save 477.12 NOK
    9% Off
In Stock
Store lab supplies that don't typically fit in standard-size compartments with this versatile, expandable system.
  • Compatible with all standard LabStack cabinets and options
  • 8-Drawer Cabinet comes with two rows of four drawers
  • Each drawer has enough dividers to produce seven rows 1.2 in. (30mm) wide
  • Accepts embedding rings, processing cassettes, and unmounted paraffin blocks
TRUSTED_SUSTAINABILITY
Specifications
Product Type 8-Drawer Cabinet
Color Gray