missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ R2A Agar (Dehydrated)

Product Code. p-10714001
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
10651655 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 10651655

Brand: Thermo Scientific™ CM0906B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enumerate heterotrophic bacteria from drinking water samples with this nutritionally reduced medium to prevent overgrowth.

Enumerate heterotrophic bacteria from drinking water samples with Thermo Scientific™ Oxoid™ R2A Agar (Dehydrated). This nutritionally reduced medium was specially designed to enhance the recovery of stressed and chlorine damaged bacteria from treated water samples. Stressed or injured organisms during water treatment are unable to grow on high nutrient media since faster-growing organisms outgrow the former. Therefore the use of a low nutrient medium like R2A Agar incubated for longer incubation periods allows these stressed organisms to grow well.

Standard methods for the enumeration of heterotrophic bacteria in water have traditionally used nutritionally rich media, such as Plate Count Agar, with incubation at 35°C1. Organisms isolated under these conditions may represent only a small percentage of the bacteria present in the sample2.

R2A Agar aids recovery of stressed and damaged bacteria giving more realistic colony count as compared to traditional media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: EP, BP, USP.
Description R2A Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 27.6L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.