missing translation for 'onlineSavingsMsg'
Learn More

R162, MedChemExpress

Product Code. p-200096081
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16486517 5 mg 5mg
16484846 100 mg 100mg
16433798 10 mg 10mg
16432866 25 mg 25mg
16430177 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16486517

Brand: Medchem Express HY103096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R162 is a potent inhibitor of glutamate dehydrogenase 1 (GDH1/GLUD1), with anti-cancer properties.

R162 is a potent inhibitor of glutamate dehydrogenase 1 (GDH1/GLUD1), with anti-cancer properties.

Specifications

Chemical Name or Material R162
CAS 64302-87-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H12O3
Quantity 5 mg
Solubility Information DMF : 6.25 mg/mL (23.65 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 5.2 mg/mL (19.68 mM; Need ultrasonic) ∣Ethanol : 2 mg/mL (7.57 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1C2=C(C=CC=C2)C(C3=CC=C(CC=C)C(O)=C13)=O
Molecular Weight (g/mol) 264.28
Formula Weight 264.28
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.