Learn More
Brady™ R10000 Series Printer Ribbon
Product Code. 17545136
Change view
Click to view available options
Color:
Black
Blue
Green
Magenta
Red
White
Yellow
Unit Size:
Each
Product Code. Color unitSize
17545136 Magenta Each
17585126 Black Each
17515136 Blue Each
17525136 Green Each
17505136 Red Each
17595126 White Each
17535136 Yellow Each
7 options
Product Code. 17545136
Brand: Brady™ 151006
Only null left
Add to basket
Add to basket
Switch out your ribbon in seconds with these easy-to-load ribbon cartridges that slide into place in the always-correct direction.
  • Make it last – R10000 series ribbons are based on a resin formulation that produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions
  • Make it fast – self-contained ribbon cartridges are easy to replace eliminating the hassle of printer calibration and ribbon threading
  • R10000 series ribbons are ideal for use with long-lasting indoor / outdoor materials such as Brady B-595
Specifications
Product Type Printer Ribbon
Color Magenta
For Use With (Equipment) BMP61 Labeling System, BradyPrinter M611
Material Resin
Print Technology Thermal Transfer
Length (English) 75 ft.
Width (English) 2 in.
Dimensions (L x W) 75 ft. x 2 in.
Alternate Part Number M61-R10010-MA