Learn More
(R)-Xanthoanthrafil, TRC
Product Code. 30457281
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457281 1 mg 1mg
30451061 10 mg 10mg
30453014 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30457281 Supplier Toronto Research Chemicals Supplier No. TRCX5000051MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 247568-68-9
Molecular Formula C19 H23 N3 O6
Molecular Weight (g/mol) 389.4
Synonym N-[(3,4-Dimethoxyphenyl)methyl]-2-[[(1R)-2-hydroxy-1-methylethyl]amino]-5-nitro-benzamide, FR 226807
IUPAC Name N-[(3,4-dimethoxyphenyl)methyl]-2-[[(2R)-1-hydroxypropan-2-yl]amino]-5-nitrobenzamide
SMILES COc1ccc(CNC(=O)c2cc(ccc2N[C@H](C)CO)[N+](=O)[O-])cc1OC
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C19H23N3O6/c1-12(11-23)21-16-6-5-14(22(25)26)9-15(16)19(24)20-10-13-4-7-17(27-2)18(8-13)28-3/h4-9,12,21,23H,10-11H2,1-3H3,(H,20,24)/t12-/m1/s1
Formula Weight 389.1587
Chemical Name or Material (R)-Xanthoanthrafil