missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 1 Antibody (Oncomed patent anti-RSPO1) [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30171021
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30171021 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30171021 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328641JF585
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Human Monoclonal Antibody

R-Spondin 1 Monoclonal antibody specifically detects R-Spondin 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen R-Spondin 1
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Oncomed patent anti-RSPO1
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Host Species Human
Immunogen RSPO1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 284654
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.