Learn More
(R,S)-N-Nitroso Anatabine-2,4,5,6-d4, TRC
Specifications
Specifications
| Chemical Name or Material | (R,S)-N-Nitroso Anatabine-2,4,5,6-d4 |
| CAS | 1020719-69-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 2H4 H7 N3 O |
| Quantity | 0.5 mg |
| Synonym | 2,3'-Bipyridine-2',4',5',6'-d4','1,2,3,6-tetrahydro-1-nitroso-, (RS)-N-Nitrosoanatabine D4 (pyridinyl D4), (R,S)-N-Nitroso Anatabine-2,4,5,6-d4, (R,S)-N-Nitrosoanatabine D4 (pyridinyl D4), 1,2,3,6-Tetrahydro-1-nitroso-2,3'-bipyridine-2',4',5',6'-d4 (ACI) |
| InChI Formula | InChI=1S/C10H11N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h1-4,6,8,10H,5,7H2/i3D,4D,6D,8D |
| SMILES | [2H]c1nc([2H])c(C2CC=CCN2N=O)c([2H])c1[2H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(1-nitroso-3,6-dihydro-2H-pyridin-2-yl)pyridine |
| Molecular Weight (g/mol) | 193.24 |
| Show More |
Product Title
¿Detecta una oportunidad de mejora?