Learn More
(R,S)-N-Nitroso Anatabine-2,4,5,6-d4, TRC
Product Code. 30480143
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480143 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30480143 Supplier Toronto Research Chemicals Supplier No. TRCN5247525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R,S)-N-Nitroso Anatabine-2,4,5,6-d4
CAS 1020719-69-0
Purity Grade Notes HPLC
Molecular Formula C10 2H4 H7 N3 O
Quantity 5 mg
Synonym 2,3'-Bipyridine-2',4',5',6'-d4, 1,2,3,6-tetrahydro-1-nitroso-, (RS)-N-Nitrosoanatabine D4 (pyridinyl D4), (R,S)-N-Nitroso Anatabine-2,4,5,6-d4, (R,S)-N-Nitrosoanatabine D4 (pyridinyl D4), 1,2,3,6-Tetrahydro-1-nitroso-2,3'-bipyridine-2',4',5',6'-d4 (ACI)
InChI Formula InChI=1S/C10H11N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h1-4,6,8,10H,5,7H2/i3D,4D,6D,8D
SMILES [2H]c1nc([2H])c(C2CC=CCN2N=O)c([2H])c1[2H]
IUPAC Name 2,3,4,6-tetradeuterio-5-(1-nitroso-3,6-dihydro-2H-pyridin-2-yl)pyridine
Molecular Weight (g/mol) 193.24
Formula Weight 193.1153
Percent Purity >95
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.