Learn More
R&D Systems™ Parameter Immunoassay Control Set 1010 for Corticosterone
Product Code. 18431180
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18431180 1 Each 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18431180 Supplier R&D Systems™ Supplier No. QC205
  • ONLINE EXCLUSIVE PRICE
    2230.00 NOK
    2105.25 NOK / 1 sets
    Save 124.75 NOK
    5% Off
 Restricted Item
Estimated Shipment: 23-11-2026
For use with catalog number KGE009
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Corticosterone
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Multi-species