Learn More
R&D Systems™ Parameter Immunoassay Control Set 594 for LTB4
Product Code. 18714988
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18714988 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18714988 Supplier R&D Systems™ Supplier No. QC116
  • ONLINE EXCLUSIVE PRICE
    2230.00 NOK
    2105.25 NOK / Pack of 1
    Save 124.75 NOK
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 24-09-2026
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
For use with catalog numbers KGE006B, SKGE006B, PKGE006B
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Leukotriene B4 (LTB4)
Intended Use Control
Quantity 1 Each
Source Multi-species