Learn More
(R)-Oxiracetam, MedChemExpress
Product Code. 16437398
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16437398
Brand: Medchem Express HY17554
MedChemExpress (R)-Oxiracetam is the (R)-enantiomer of the nootropic drug oxiracetam. Oxiracetam (ISF 2522) is a nootropic drug of the racetam family and stimulant.
(R)-Oxiracetam is the (R)-enantiomer of the nootropic drug oxiracetam. Oxiracetam (ISF 2522) is a nootropic drug of the racetam family and stimulant.
Specifications
Chemical Name or Material (R)-Oxiracetam
CAS 68252-28-8
Color White
Purity Grade Notes Research
Molecular Formula C6H10N2O3
Quantity 10 mg
Synonym (R)-(+)-Oxiracetam
Solubility Information H2O : ≥ 50 mg/mL (316.14 mM)
SMILES O=C(N)CN1C(C[C@@H](O)C1)=O
Molecular Weight (g/mol) 158.16
Formula Weight 158.16
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.