missing translation for 'onlineSavingsMsg'
Learn More

(R)-GNE-140, MedChemExpress

Product Code. 16490689
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16490689 1 mg 1mg
16483148 10 mg 10mg
16419706 50 mg 50mg
16414446 100 mg 100mg
16435887 5 mg 5mg
5 options
This item is not returnable. View return policy

Product Code. 16490689

Brand: Medchem Express HY100742A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress (R)-GNE-140 is a potent lactate dehydrogenase A (LDHA) inhibitor, with IC50s of 3 nM and 5 nM for LDHA and LDHB, respectively; (R)-GNE-140 is 18-fold more potent than S enantiomer.

(R)-GNE-140 is a potent lactate dehydrogenase A (LDHA) inhibitor, with IC50s of 3 nM and 5 nM for LDHA and LDHB, respectively

Specifications

Chemical Name or Material (R)-GNE-140
Physical Form Solid
CAS 2003234-63-5
Color Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C25H23ClN2O3S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (100.19 mM)
SMILES O=C1N[C@@](C2=CSC=C2)(C3=CC=C(N4CCOCC4)C=C3)CC(O)=C1SC5=CC=CC=C5Cl
Molecular Weight (g/mol) 499.04
Formula Weight 499.04
Percent Purity 99.2%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.