Learn More
R&D Systems™ StemXVivo Osteogenic/Adipogenic Base Media
Product Code. 18431550
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18431550 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18431550 Supplier R&D Systems™ Supplier No. CCM007
  • ONLINE EXCLUSIVE PRICE
    2335.00 NOK
    2206.05 NOK / 250mL
    Save 128.95 NOK
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Base media for the differentiation of MSCs into osteocytes or adipocytes
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into osteocytes or adipocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
  • Supports reproducible human, mouse, and rat MSC differentiation
  • Offers flexibility to evaluate novel cytokine and growth factor combinations to induce osteogenesis or adipogenesis
  • Has been developed and optimized using MSCs
  • Can be used with StemXVivo Osteogenic Supplement (Mfr. Nos. CCM008 or CCM009) or StemXVivo Adipogenic Supplement (Mfr. Nos. CCM011) to reduce variation in differentiation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stem Cell Media
Quantity 250 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.