Learn More
R&D Systems™ MagCellect Rat B Cell Isolation Kit
Product Code. 18431270
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18431270 1 Kit 1 set
1 options
Product Code. 18431270
Brand: R&D Systems™ MAGR303
Designed to isolate B cells via a negative selection principle
Typical recovery of the targeted cell population ranges between 40% and 65% and the purity of recovered B cells ranges from 89-95%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
Magnetically tagged cells will migrate toward the magnet (unwanted cell fraction)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit