Learn More
R&D Systems™ MagCellect Mouse CD8+ T Cell Isolation Kit
Product Code. 18472600
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18472600 1 Kit 1 set
1 options
Product Code. 18472600
Brand: R&D Systems™ MAGM203
Designed to isolate CD8+ T cells via a negative selection principle
CD8 is a heterodimeric glycoprotein consisting of an alpha and beta chain. It is expressed on cytolytic T cells and functions in conjunction with the T cell receptor in the recognition of MHC/peptide complexes.
Typical recovery ranges between 40 and 55% and the purity of recovered CD8+ T cells ranges between 87 and 93%, with no detectable CD4+ T cells
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit