Learn More
R&D Systems™ MagCellect Human Naive CD4+ T Cell Isolation Kit
Product Code. 18421660
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Kit
missing translation for 'unitSize'
1 set
Product Code. 18421660
missing translation for 'mfr': R&D Systems™ MAGH115
Designed to isolate CD4+ Naive T Cells via a negative selection principle
CD4 is an approximately 55kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T cells. In humans, CD4 is also expressed to a lesser extent on monocytes and macrophage related cells.
Typical purity of recovered CD4+ Naive T Cells ranges between 75% and 85%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit