Learn More
R&D Systems™ MagCellect Human B Cell Isolation Kit
Product Code. 18461690
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18461690 1 Kit 1 set
1 options
Product Code. 18461690
Brand: R&D Systems™ MAGH103
Designed to isolate B cells via a negative selection principle
The resulting cell preparation is highly enriched with B cells
Typical recovery of the targeted cell population ranges between 45% and 65%
Purity of recovered B cells ranges between 90 and 98%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit