Learn More
R&D Systems™ DLDVPIPGRFDRRVS(PO3)VAAE Ser/Thr Phosphatase Substrate I
Product Code. 18349687
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349687 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18349687 Supplier R&D Systems™ Supplier No. ES012
  • ONLINE EXCLUSIVE PRICE
    3275.00 NOK
    3097.50 NOK / 1mg
    Save 177.50 NOK
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
Phosphopeptide substrate for Calcineurin, PP1, PP2A, PP2C, and Ser/Thr phosphatases
  • Synthetic phosphopeptide corresponding to amino acids 81 to 99 of Protein Kinase A Regulatory Subunit 2-alpha
  • Enzymatic cleavage of this substrate liberates inorganic phosphate, which can be measured in dye binding assays, such as the malachite green method
  • Purity 95% based on HPLC analysis
TRUSTED_SUSTAINABILITY
Specifications
Form Lyophilized
Quantity 1 mg
Format Lyophilized from a stock solution in distilled water.
Molecular Weight (g/mol) Theoretical Molecular weight: 2192 Da
Product Type Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-Arg-Val-Ser(PO3)-Val-Ala-Ala-Glu
Storage Requirements No Stability Data Storage is set to N/A for ProdCode