Learn More
R&D Systems™ CellXVivo Human Monocyte-derived DC Differentiation Kit
For the differentiation of monocyte-derived dendritic cells
Brand:  R&D Systems™ CDK004
Encompass
Product Code. 15720073
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Serum-Free Dendritic Cell Base Media; Recombinant Human IL-4; Recombinant Human GM-CSF; Recombinant Human TNF-alpha; Reconstitution Buffer 2
Description
Description
For the differentiation of CD14+ monocytes into dendritic cells.