Learn More
R&D Systems™ CellXVivo Human Monocyte-derived DC Differentiation Kit
For the differentiation of monocyte-derived dendritic cells
Brand: R&D Systems™ CDK004
Includes: Serum-Free Dendritic Cell Base Media; Recombinant Human IL-4; Recombinant Human GM-CSF; Recombinant Human TNF-alpha; Reconstitution Buffer 2
Description
For the differentiation of CD14+ monocytes into dendritic cells.