Learn More
(R)-(+)-Cysteine, Sigma-Aldrich™
Product Code. 15860211
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860211 100 g 100g
15826550 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15860211 Supplier Sigma-Aldrich Supplier No. 8160140100
 Restricted Item
Estimated Shipment: 12-11-2026
For synthesis
Cysteine is used as a building block in the synthesis of peptides due to its ability to form disulfide bonds. Additionally, it functions as a chiral auxiliary in asymmetric synthesis, facilitating the induction of chirality in the resulting products.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52-90-4
Grade Research
Molecular Weight (g/mol) 121.16
Specifications
Melting Point 220°C to 228°C
MDL Number MFCD00064306
Synonym (R)-(+)-Cysteine, α-Amino-β-mercapto propionic acid, αlpha-Amino-β-mercapto propionic acid, Cys, αlpha-Amino-beta-mercapto propionic acid, (R)-(+)-Cysteine
Quantity 100 g
Chemical Name or Material (R)-(+)-Cysteine