missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-beta-Hydroxy-gamma-butyrolactone, 90+%, ee 99+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(R)-(+)-β-Hydroxy-γ-butyrolactone is used in the pharmaceutical industry as a chiral building block for the statin class of cholesterol-reducing drugs. It is also used to prepare HIV inhibitors and nutritional supplement such as L-carnitine.
Solubility
Miscible with ethanol and chloroform.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
(R)-(+)-β-Hydroxy-γ-butyrolactone is used in the pharmaceutical industry as a chiral building block for the statin class of cholesterol-reducing drugs. It is also used to prepare HIV inhibitors and nutritional supplement such as L-carnitine.
Solubility
Miscible with ethanol and chloroform.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Density | 1.302 |
Boiling Point | 98°C to 100°C (0.3 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.4655 |
Quantity | 5 g |
Beilstein | 3648447 |
Solubility Information | Miscible with ethanol and chloroform. |
Optical Rotation | +81° (c=2 in Methanol) |
Formula Weight | 102.09 |
Percent Purity | ≥90% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'