missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Methylbenzylamine, TRC

Product Code. 17991025
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991025 1 g 1g
30134555 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991025 Supplier Toronto Research Chemicals Supplier No. TRCM2888001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3886-69-9
InChI Formula InChI=1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m1/s1
IUPAC Name (1R)-1-phenylethanamine
Molecular Formula C8 H11 N
Molecular Weight (g/mol) 121.18
Chemical Name or Material (R)-(+)-a-Methylbenzylamine

Specifications

Quantity 1 g
Synonym Benzenemethanamine, α-methyl-, (αR)-, Benzenemethanamine, α-methyl-, (R)-, Benzylamine, α-methyl-, (R)-(+)- (8CI), Benzylamine, α-methyl-, d- (4CI), (αR)-α-Methylbenzenemethanamine, (+)-(R)-1-Phenethylamine, (+)-(R)-1-Phenylethylamine, (+)-(R)-α-Methylbenzylamine, (+)-(R)-α-Phenylethylamine, (+)-1-Amino-1-phenylethane, (+)-1-Phenethylamine, (+)-1-Phenylethan-1-amine, (+)-1-Phenylethylamine, (+)-Phenylethylamine, (+)-[(1R)-1-Phenylethyl]amine, (+)-α-Methylbenzenemethanamine, (+)-α-Methylbenzylamine, (+)-α-Phenethylamine, (+)-α-Phenylethylamine, (1R)-1-Phenyl-1-ethanamine, (1R)-1-Phenylethan-1-amine, (1R)-1-Phenylethanamine, (1R)-1-Phenylethanamine, (1R)-1-Phenylethylamine, (1R)-Phenylethylamine, (R)-(+)-1-Methylbenzylamine, (R)-(+)-1-Phenethylamine, (R)-(+)-1-Phenylethanamine, (R)-(+)-1-Phenylethylamine, (R)-(+)-Alpha-methylbenzylamine, (R)-(+)-Phenethylamine, (R)-(+)-α-Methylbenzylamine, (R)-(+)-α-Phenethylamine, (R)-(+)-α-Phenylethylamine, (R)-1-Amino-1-phenylethane, (R)-1-Aminoethylbenzene, (R)-1-Phenethylamine, (R)-1-Phenyl-1-ethanamine, (R)-1-Phenylethanamine, (R)-1-Phenylethylamine, (R)-Alpha-methylbenzylamine, (R)-Methylbenzylamine, (R)-Phenethylamine, (R)-Phenylethylamine, (R)-Phenylethylamine, (R)-α-Aminoethylbenzene, (R)-α-Methylbenzenemethanamine, (R)-α-Methylbenzylamine, (R)-α-Phenethylamine, (R)-α-Phenylethylamine, D-(+)-1-Phenylethylamine, D-(+)-α-Methylbenzylamine, D-(+)-α-Phenylethylamine, D-1-Phenylethylamine, D-Phenethylamine, D-α-Methylbenzylamine, D-α-Phenethylamine, R-(+)-Methylbenzylamine, R-(+)-Phenylethylamine, d-1-Phenylethylamine, Levetiracetam Imp. E (EP)
SMILES C[C@@H](N)c1ccccc1
Formula Weight 121.0891
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.