Learn More
R 428, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(6,7-Dihydro-5H-benzo[6,7]cyclohepta[1,2-c]pyridazin-3-yl)-N3-((7-(S)-pyrrolidin-1-yl)-6,7,8,9-tetrahydro-5H-benzo[7]annulene-2-yl)-1H-1,2,4-triazole-3,5-diamine |
| CAS | 1037624-75-1 |
| Molecular Formula | C30H34N8 |
| Quantity | 10 mg |
| Synonym | BGB 324, R 428 |
| SMILES | Nc1nc(Nc2ccc3CC[C@@H](CCc3c2)N4CCCC4)nn1c5cc6CCCc7ccccc7c6nn5 |
| IUPAC Name | 1-(6,7-dihydro-5H-benzo[4,5]cyclohepta[1,2-d]pyridazin-3-yl)-3-N-[(7S)-7-pyrrolidin-1-yl-6,7,8,9-tetrahydro-5H-benzo[7]annulen-3-yl]-1,2,4-triazole-3,5-diamine |
| Molecular Weight (g/mol) | 506.64 |
| Formula Weight | 506.2906 |