missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Aminomethyl-1-Boc-piperidine, 97%, Thermo Scientific Chemicals

Catalog Number p-7034861
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
140645-23-4
C11H22N2O2
214.309
MFCD03839876
WPWXYQIMXTUMJB-SECBINFHSA-N
r-1-boc-3-aminomethyl piperidine, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, r-1-n-boc-3-aminomethyl piperidine, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine
1502023
tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CN
This item is not returnable. View return policy
140645-23-4
C11H22N2O2
214.309
MFCD03839876
WPWXYQIMXTUMJB-SECBINFHSA-N
r-1-boc-3-aminomethyl piperidine, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, r-1-n-boc-3-aminomethyl piperidine, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine
1502023
tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CN

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material (R)-3-Aminomethyl-1-Boc-piperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'