missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 95%, Thermo Scientific™

Product Code. 15427396
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15427396 1 g 1g
15447396 250 mg 250mg
15437396 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15427396

Brand: Thermo Scientific Alfa Aesar H62403.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131570-56-4
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00236844
InChI Key MVWPBNQGEGBGRF-LJQANCHMSA-N
Synonym boc-d-dap fmoc-oh, boc-n3-fmoc-d-2,3-diaminopropionic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-n'-fmoc-d-2,3-diaminopropionic acid, boc-d-dpr fmoc-oh, ambotzbaa1031, boc-d-dapa fmoc-oh, pubchem14750, rarechem bk pt 0245, boc-d-dapa fmoc-oh.solvent
PubChem CID 2755948
IUPAC Name (2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 426.46
Percent Purity 95%
Chemical Name or Material (R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.