Learn More
(R)-(+)-2-Acetoxypropionyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-(+)-2-Acetoxypropionyl Chloride |
| CAS | 53636-19-4 |
| Molecular Formula | C5H7ClO3 |
| Quantity | 500 mg |
| Synonym | Propanoyl chloride','2-(acetyloxy)-','(2R)- (9CI','ACI), (2R)-2-(Acetyloxy)propanoyl chloride (ACI), Propanoyl chloride','2-(acetyloxy)-','(R)- (ZCI), (2R)-1-Chloro-1-oxopropan-2-yl acetate, (R)-2-Acetoxypropanoyl chloride, (R)-2-Acetoxypropionyl chloride, 1-(R)-Chlorocarbonylethyl acetate, D-O-Acetyllactoyl chloride, D-α-Acetoxypropionyl chloride, O-Acetyl-D-lactoyl chloride |
| InChI Formula | InChI=1S/C5H7ClO3/c1-3(5(6)8)9-4(2)7/h3H,1-2H3/t3-/m1/s1 |
| SMILES | [C@H](OC(C)=O)(C(Cl)=O)C |
| IUPAC Name | (R)-1-chloro-1-oxopropan-2-yl acetate |
| Molecular Weight (g/mol) | 150.56 |
| Formula Weight | 150.01 |