Learn More
(R)-1-Phenylpropan-1-amine, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 3082-64-2 |
| Molecular Formula | C9H13N |
| MDL Number | MFCD00083057 |
| Quantity | 500 g |
| SMILES | N[C@H](CC)C1=CC=CC=C1 |
| Molecular Weight (g/mol) | 135.21 |
| Percent Purity | 95% |
| Chemical Name or Material | (R)-1-Phenylpropan-1-amine |