missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenyl-1-propanol, 99%

Product Code. p-7044289
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357545 100 mg 100mg
11347545 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357545 Supplier Thermo Scientific Alfa Aesar Supplier No. L05681.MC
Add to basket
This item is not returnable. View return policy

CAS: 1565-74-8 | C9H12O | 136.194 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1565-74-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00064279
InChI Key DYUQAZSOFZSPHD-SECBINFHSA-N
Synonym r-+-1-phenyl-1-propanol, r-1-phenylpropan-1-ol, 1r-1-phenylpropan-1-ol, r-1-phenyl-1-propanol, r-1-phenyl-propanol, r-+-alpha-ethylbenzyl alcohol, r-+-1-phenylpropanol, r-1-phenylpropanol, 1-phenyl-1-propanol #, r-1-phenyl-1propanol
PubChem CID 640199
IUPAC Name (1R)-1-phenylpropan-1-ol
SMILES CCC(C1=CC=CC=C1)O

Specifications

Density 0.993
Boiling Point 217°C to 219°C
Flash Point 95°C (203°F)
Refractive Index 1.52
Quantity 100 mg
Beilstein 2041555
Optical Rotation +47° (c=2.2 in Hexane)
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material (R)-(+)-1-Phenyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.