missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-BOC-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific™

Catalog Number p-4228392
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
140695-85-8
C11H21NO3
215.29
OJCLHERKFHHUTB-SECBINFHSA-N
r-1-boc-3-hydroxymethyl piperidine, r-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, r-1-boc-3-hyroxymethyl piperidine, tert-butyl 3r-3-hydroxymethyl piperidine-1-carboxylate, r-n-boc-3-hydroxymethyl piperidine, r-n-boc-3-piperidinemethanol, r-1-n-boc-3-hydroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl-piperidine, r---1-boc-3-hydroxymethyl piperidine
1514447
tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CO
This item is not returnable. View return policy
140695-85-8
C11H21NO3
215.29
OJCLHERKFHHUTB-SECBINFHSA-N
r-1-boc-3-hydroxymethyl piperidine, r-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, r-1-boc-3-hyroxymethyl piperidine, tert-butyl 3r-3-hydroxymethyl piperidine-1-carboxylate, r-n-boc-3-hydroxymethyl piperidine, r-n-boc-3-piperidinemethanol, r-1-n-boc-3-hydroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl-piperidine, r---1-boc-3-hydroxymethyl piperidine
1514447
tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CO
CAS Min % 96.0
CAS Max % 100.0
Melting Point 92°C to 93°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -11.2
Formula Weight 215.29
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-1-BOC-3-(Hydroxymethyl)piperidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'