Learn More
(R)-(+)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 98%, Thermo Scientific™
Catalog Number p-7047403
Click to view available options
Quantity:
5 g
25 g
374898-01-8
C8H10FN
139.173
MFCD03093090
QGCLEUGNYRXBMZ-ZCFIWIBFSA-N
r-1-4-fluorophenyl ethanamine, r-1-4-fluorophenyl ethylamine, 1r-1-4-fluorophenyl ethanamine, r-1-4-fluorophenyl-ethylamine, r-+-1-4-fluorophenyl ethylamine, 1r-+-1-4-fluorophenyl ethylamine, pubchem15219, 1r-1-4-fluorophenyl ethylamine, r-1-4-fluoro-phenyl ethylamine, 4-fluoro-alpha-r-methylbenzylamine
6950187
(1R)-1-(4-fluorophenyl)ethanamine
CC(C1=CC=C(C=C1)F)N
374898-01-8
C8H10FN
139.173
MFCD03093090
QGCLEUGNYRXBMZ-ZCFIWIBFSA-N
r-1-4-fluorophenyl ethanamine, r-1-4-fluorophenyl ethylamine, 1r-1-4-fluorophenyl ethanamine, r-1-4-fluorophenyl-ethylamine, r-+-1-4-fluorophenyl ethylamine, 1r-+-1-4-fluorophenyl ethylamine, pubchem15219, 1r-1-4-fluorophenyl ethylamine, r-1-4-fluoro-phenyl ethylamine, 4-fluoro-alpha-r-methylbenzylamine
6950187
(1R)-1-(4-fluorophenyl)ethanamine
CC(C1=CC=C(C=C1)F)N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(R)-(+)-1-(4-Fluorophenyl)ethylamine, is used as a pharmaceutical intermediate and as a chemical in organic synthesis
Solubility
Sparingly Soluble in water (1.5E-3 g/L) (25°C).
Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Melting Point -30°C
Density 1.03
Boiling Point 76°C (22 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.502
Quantity 5 g
UN Number UN2735
Beilstein 8614058
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble in water (1.5E-3g/L) (25°C).
Optical Rotation +33° (Neat)
Formula Weight 139.17
Percent Purity 99%
Assay ee 98%
Chemical Name or Material (R)-(+)-1-(4-Fluorophenyl)ethylamine