missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 98+%, Thermo Scientific™

Product Code. 11323916
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323916 5 g 5g
11313916 1 g 1g
11333916 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323916 Supplier Thermo Scientific Alfa Aesar Supplier No. L17021.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

Solubility
Soluble in water (6.91g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23357-46-2
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.221
MDL Number MFCD00671629
InChI Key JRZGPXSSNPTNMA-SNVBAGLBSA-N
Synonym r-1,2,3,4-tetrahydronaphthalen-1-amine, r---1,2,3,4-tetrahydro-1-naphthylamine, 1r-1,2,3,4-tetrahydronaphthalen-1-amine, r-1,2,3,4-tetrahydro-1-naphthylamine, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1r, r-1-amino-1,2,3,4-tetrahydronaphthalene, r-1, 2, 3, 4-tetrahydro-1-naphthylamine, r-1-aminotetraline, tetralin-1alpha-amine, r-1-aminotetralin
PubChem CID 7058072
IUPAC Name (1R)-1,2,3,4-tetrahydronaphthalen-1-amine
SMILES C1CC(C2=CC=CC=C2C1)N

Specifications

Density 1.035
Boiling Point 118°C to 120°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5645
Quantity 5 g
Beilstein 4231302
Sensitivity Air Sensitive
Solubility Information Soluble in water (6.91g/L at 25°C).
Formula Weight 147.22
Percent Purity ≥99%
Assay ee ≥98%
Chemical Name or Material (R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.