Learn More
(R)-(+)-1,1′-Binaphthyl-2,2′-diol, Sigma-Aldrich™
Product Code. 15830161
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15830161 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15830161 Supplier Sigma Aldrich Supplier No. 8147740010
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18531-94-7
Grade Research
Molecular Weight (g/mol) 286.32
Specifications
Melting Point 203°C to 207°C
MDL Number MFCD00004068
Quantity 10 g
Synonym (R)-(+)-1,1′-Binaphthyl-2,2′-diol, (R)-(+)-2,2′-Dihydroxy-1,1′-binaphthyl, (R)-(+)-1,1′-Binaphthyl-2,2′-diol