Få mere at vide
Få mere at vide
Quinoxaline-2-carboxylic acid-d4, Honeywell Fluka™
Shop alle Honeywell Fluka produkter
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
Förpackningsstorlek
10mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
| Packaging | Glass Bottle |
| Physical Form | Solid |
| Industry Type | Antibacterial Agents |
| Quantity | 10 mg |
| Grade | Analytical Standard |