Learn More
Quinoline, Sigma-Aldrich™
Product Code. 32237626
Change view
Click to view available options
Quantity:
49 kg
Unit Size:
49kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32237626 49 kg 49kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32237626 Supplier Sigma Aldrich Supplier No. 8024079049
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Quinoline
CAS 91-22-5
Boiling Range 237°C to 238°C/1013 hPa
Melting Point -15°C
Density 1.09 g/cm³ at 25°C
Flash Point 107°C (closed cup)
Quantity 49 kg
Synonym Quinoline, 1-Azanaphthalene, Quinoline
Molecular Weight (g/mol) 129.16
Grade Not Available