Learn More
Quinoline-3-boronic acid, 95%
Product Code. 11344526
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
11344526 1 g 1g
11364526 250 mg 250mg
11354526 5 g 5g
3 options
Product Code. 11344526
Brand: Thermo Scientific Alfa Aesar L20088.03
Request Bulk or Custom Format
CAS: 191162-39-7 | C9H8BNO2 | 172.98 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 191162-39-7
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.98
MDL Number MFCD02183527
InChI Key YGDICLRMNDWZAK-UHFFFAOYSA-N
Synonym 3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl
PubChem CID 2734663
SMILES OB(O)C1=CC2=CC=CC=C2N=C1
Specifications
Melting Point >300°C
Quantity 1 g
Beilstein 8764547
IUPAC Name (quinolin-3-yl)boronic acid
Formula Weight 172.98
Percent Purity 95%
Chemical Name or Material Quinoline-3-boronic acid