Description
Clean interior surfaces, tools, or products. Quiltec™ I Wipes offer an extremely high sorptive capacity. These durable polyester wipes are ideal for cleanroom applications.
- These sealed-edge wipes offer an extremely high sorptive capacity. Two layers of quilted polyester fabric are laser-cut and sealed together to create pockets where additional fluid can be trapped.
- Two-ply quilted polyester construction creates durable wiper with exceptional absorbency—absorbs more than twice the capacity of a single-layer wiper
- Extremely low in both particles and fibers—ideal for interior surface cleaning of process tools, wiping down products, or general-purpose cleaning
Order Info
Laundered and packaged in an ISO Class 4 cleanroom.
Specifications
Specifications
| Product Type | Cleanroom Wipe |
| Material | Polyester |
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | ISO Class 3 and 4 |
| Disposable | Single-use |
| Dimensions (L x W) | 23 x 23 cm (9 x 9 in.) |
| For Use With (Application) | Cleanroom |
| Packaging Quantity | 25/Bag, 40 Bags/Cs. |
| Ply | 2-ply |
| Sterility | Sterile |