Learn More
Quillaic acid, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Quillaic acid(Quillaja sapogenin) is the major aglycone of the widely studied saponins of the Chilean indigenous tree Quillaja saponaria Mol
Specifications
Specifications
| Chemical Name or Material | Quillaic acid |
| CAS | 631-01-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C30H46O5 |
| Quantity | 5 mg |
| Synonym | Quillaja sapogenin |
| Solubility Information | DMSO : ≥ 41 mg/mL (84.24 mM) |
| SMILES | OC([C@]1(CCC(C)(C)C2)[C@]2([H])C3=CC[C@@]4([H])[C@@](C)(CC[C@]5([H])[C@@]4(CC[C@H](O)[C@@]5(C)C=O)C)[C@]3(C)C[C@H]1O)=O |
| Molecular Weight (g/mol) | 486.68 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.