Learn More
Quickfit™ Screwthread Vacuum Receiver Adapter
Product Code. 15507950
Click to view available options
Material:
Borosilicate Glass
Unit Size:
Pack of 5
Product Code. 15507950
Brand: Quickfit RA11/SC
Only null left
Add to basket
Add to basket
Allows four flasks to be connected to a condenser outlet.
  • Useful for collecting fractions when distilling under vacuum
  • Each flask can be progressively filled by rotating the adapter
  • Avoids problems of disturbing vacuum conditions when changing flasks
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Useful for collecting distillate fractions
For Use With (Equipment) Flask
Includes Screw thread connector
Material Borosilicate Glass
Product Type Vacuum receiver adapter