Learn More
Quickfit™ Unbadged Joints Cones
Product Code. 15507770
Change view
Click to view available options
Cone Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
40/38
45/40
55/44
7/16
Thickness (Metric) Wall:
1mm
1.5mm
1.8mm
2.00mm
2mm
2.30mm
5mm
Unit Size:
Pack of 10
Pack of 20
Pack of 5
Pack of 50
Product Code. Cone Size Thickness (Metric) Wall unitSize
15507770 55/44 5mm Pack of 20
12939363 10/19 1mm Pack of 10
12468816 14/23 1.5mm Pack of 10
10830251 19/26 1.8mm Pack of 10
12614911 29/32 2mm Pack of 10
12488816 24/29 1.8mm Pack of 10
12458826 7/16 1.5mm Pack of 10
10735671 40/38 2mm Pack of 5
12653012 45/40 2.30mm Pack of 5
13431829 34/35 2.00mm Pack of 10
15597760 12/21 1.5mm Pack of 50
11 options
Product Code. 15507770
Brand: Quickfit CNB55UB
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Free from any ceramic marking
  • Avoids problem of badge charring at high temperatures which can give an untidy appearance
  • Manufactured to accuracy which surpasses requirements of ISO 383
  • Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
  • Cones manufactured with shank length of 100mm
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5Pack
Type Male Ground Joint
Cone Size 55/44
Thickness (Metric) Wall 5mm