Learn More
Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Product Code. 12508875
Click to view available options
Standard Taper Joint:
7/16
Standard; 10/19
Standard; 12/21
Standard; 14/23
Standard; 19/26
Standard; 24/29
Standard; 29/32
Standard; 34/35
Standard; 45/40
Unit Size:
Pack of 10
Pack of 2
Pack of 5
Product Code. 12508875
Brand: Quickfit SB29
Only null left
Add to basket
Add to basket
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. Azlon™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. AzlonTM Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
  • Conforms to the standard tapers as outlined in ISO 383
  • Borosilicate glass for chemical durability and high temperature performance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chemical durability and high temperature performance
For Use With (Equipment) Quickfit™ jointed ware and other Glassware having standard ground Glass sockets
Standard Taper Joint Standard; 29/32
Material Borosilicate Glass
Product Type Hollow Stopper