Learn More
Quickfit™ Right Angle Tubing Adapter
Description
Quickfit™ Right Angle Tubing Adapter are available in a variety of cone sizes.
- With right angle connection for attachment of rubber hose etc
- Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
Specifications
Specifications
| Cone Size | 24/29 |
| For Use With (Equipment) | Flexible tubing, Rubber hose |
| Includes | Detachable Plastic screw thread connector |
| Material | Borosilicate Glass |
| Product Type | Right angle tubing adapter |