Learn More
Quickfit™ Reduction Socket Adapter
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
- Useful for the connection of dissimilar sized ground glass joints
- Small sized socket at the top, large sized cone at the bottom
Specifications
Specifications
| Cone Size | Cone Size: 45/40 |
| For Use With (Equipment) | Dissimilar sized ground Glass joint |
| Includes | Small socket, large cone |
| Material | Borosilicate Glass |
| Product Type | Reduction adapter |
| Standard Taper Joint No. | 29/32 |