Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
Brand: Quickfit DA45
776.82 NOK valid until 2025-10-26
Use promo code "24136" to get your promotional price.
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
- Useful for the connection of dissimilar sized ground glass joints
- Small sized socket at the top, large sized cone at the bottom
Specifications
Cone Size: 34/35 | |
Small socket, large cone | |
Reduction adapter |
Dissimilar sized ground Glass joint | |
Borosilicate Glass | |
29/32 |
Promotions
-
Multiple Manufacturers
Focus on Agri-Science Testing
Use promotion code 24136 to get amazing savings.Promo Code: 24136Expires: 2025-10-26
New Offer Save Up to 87%