Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
Brand:  Quickfit DA24
364.73 NOK valid until 2025-10-26
Use promo code "24136" to get your promotional price.
 View more versions of this product
Product Code. 10765861
  • 455.00 NOK / Pack of 2
Focus on Agri-Science Testing
Use promo code 24136 at checkout
View Available Promotions
Only null left
Estimated Shipment: 13-11-2025
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
  • Useful for the connection of dissimilar sized ground glass joints
  • Small sized socket at the top, large sized cone at the bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cone Size: 29/32
Small socket, large cone
Reduction adapter
Dissimilar sized ground Glass joint
Borosilicate Glass
19/26
Documents
Product Information
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Use promotion code 24136 to get amazing savings.
    Promo Code: 24136
    Expires2025-10-26
    Days
    16
    Hours
    23
    Minutes
    40
    Seconds
    28
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24158
    Expires2025-10-31
    Days
    21
    Hours
    23
    Minutes
    40
    Seconds
    28
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/09/2025 and 31/10/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Quickfit™ Reduction Socket Adapter > Cone Size: 29/32; Socket Size: 19/26